siRNA Details
| Gene Symbol: Sfpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017658.1 | - | 11 | |||
| AK141302.1 | - | 1 | 1305 | |||
| BC033603.1 | - | 1 | 2703 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sfpq| Gene Symbol: Sfpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017658.1 | - | 11 | |||
| AK141302.1 | - | 1 | 1305 | |||
| BC033603.1 | - | 1 | 2703 | |||