siRNA Details
| Gene Symbol: 9030622O22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018565.1 | - | 7 | 1011 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9030622O22Rik| Gene Symbol: 9030622O22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018565.1 | - | 7 | 1011 | ||