siRNA Details
| Gene Symbol: 9530014B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020556.1 | - | 3 | 702 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DV061028.1 | - | 3 | 741 | Chr. 13 - 53635894 - 53639191 [-] on Build GRCm38 | |

9530014B07Rik
Genomic Map