siRNA Details
| Gene Symbol: 9530014B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA671975.1 | - | 2 | 113 | ||
| AK020556.1 | - | 2 | 108 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9530014B07Rik| Gene Symbol: 9530014B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA671975.1 | - | 2 | 113 | ||
| AK020556.1 | - | 2 | 108 | |||