siRNA Details
| Gene Symbol: A430106A12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079275.1 | - | 1 | 1061 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CX633015.1 | - | 1 | 256 | Chr. 11 - 61879095 - 61879669 [-] on Build GRCm38 | |

A430106A12Rik
Genomic Map