siRNA Details
| Gene Symbol: LOC102636700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK134928.1 | - | 2 | 1055 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102636700| Gene Symbol: LOC102636700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK134928.1 | - | 2 | 1055 | ||