siRNA Details
| Gene Symbol: Gm11832 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050144.1 | - | 2 | 131 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB653715.1 | - | 2 | 131 | Chr. 4 - 11611138 - 11615007 [-] on Build GRCm38 | |

Gm11832
Genomic Map