siRNA Details
| Gene Symbol: Gm29571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050149.1 | - | 4 | 1252 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm29571| Gene Symbol: Gm29571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050149.1 | - | 4 | 1252 | ||