siRNA Details
| Gene Symbol: Gm29571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050149.1 | - | 1 | 102 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB653722.1 | - | 1 | 102 | Chr. 10 - 25536386 - 25672116 [+] on Build GRCm38 | |

Gm29571
Genomic Map