siRNA Details
| Gene Symbol: 1700019G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040255.1 | 3 | 283 | |||
| GenBank mRNA | AK006121.1 | - | 3 | 250 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700019G24Rik| Gene Symbol: 1700019G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040255.1 | 3 | 283 | |||
| GenBank mRNA | AK006121.1 | - | 3 | 250 | ||