siRNA Details
| Gene Symbol: Gm14487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK146369.1 | - | 1 | 275 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY738398.1 | - | 1 | 275 | Chr. 2 - 30534729 - 30538428 [+] on Build GRCm38 | |

Gm14487
Genomic Map