siRNA Details
| Gene Symbol: B230104C14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020954.1 | - | 1 | 596 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DT904098.1 | - | 1 | 131 | Chr. 2 - 76389029 - 76389486 [+] on Build GRCm38 | |

B230104C14Rik
Genomic Map