siRNA Details
| Gene Symbol: Gm10619 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_077222.1 | 2 | 597 | |||
| GenBank mRNA | AK030117.1 | - | 456 | |||
| AK133252.1 | - | 2 | 600 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB616852.1 | - | 4 | 456 | Chr. 7 - 73784069 - 73816431 [-] on Build GRCm38 | |

Gm10619
Genomic Map