siRNA Details
| Gene Symbol: Lemd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032278.1 | - | 1 | 801 | ||
| BC066109.1 | - | 2 | 1919 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lemd1| Gene Symbol: Lemd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032278.1 | - | 1 | 801 | ||
| BC066109.1 | - | 2 | 1919 | |||