siRNA Details
| Gene Symbol: 4930407I02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019572.1 | - | 2 | 203 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714649.1 | - | 2 | 203 | Chr. 6 - 143232337 - 143233356 [-] on Build GRCm38 | |

4930407I02Rik
Genomic Map