siRNA Details
| Gene Symbol: 4930407I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019573.1 | - | 4 | 538 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714650.1 | - | 4 | 538 | Chr. 10 - 66145296 - 66173260 [+] on Build GRCm38 | |

4930407I19Rik
Genomic Map