siRNA Details
| Gene Symbol: Particl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046011.1 | 1 | 1858 | |||
| GenBank mRNA | AK019577.1 | - | 1 | 442 | ||
| AK145188.1 | - | 1 | 1491 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX511523.1 | - | 1 | 457 | Chr. 6 - 72440086 - 72440598 [+] on Build GRCm38 | |

Particl
Genomic Map