siRNA Details
| Gene Symbol: 4930447N08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045168.1 | 3 | 602 | |||
| GenBank mRNA | AK015409.1 | - | 3 | 604 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930447N08Rik| Gene Symbol: 4930447N08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045168.1 | 3 | 602 | |||
| GenBank mRNA | AK015409.1 | - | 3 | 604 | ||