siRNA Details
| Gene Symbol: B930092H01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045334.1 | 5 | 541 | |||
| GenBank mRNA | AK047575.1 | - | 5 | 542 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B930092H01Rik| Gene Symbol: B930092H01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045334.1 | 5 | 541 | |||
| GenBank mRNA | AK047575.1 | - | 5 | 542 | ||