siRNA Details
| Gene Symbol: Il6st | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC049887.1 | - | 1464 | |||
| BC058551.1 | - | 1588 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AW323389.1 | - | 1 | 440 | Chr. 13 - 112506993 - 112507467 [+] on Build GRCm38 | |

Il6st
Genomic Map