siRNA Details
| Gene Symbol: Gm39182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038453.1 | - | 102 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB632202.1 | - | 1, 2 | 99 | Chr. 8 - 57699776 - 57727155 [+] on Build GRCm38 | |

Gm39182
Genomic Map