siRNA Details
| Gene Symbol: LOC101055656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC086661.1 | - | 2 | 706 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV365681.2 | - | 1 | 321 | Chr. 11 - 19010509 - 19011181 [+] on Build GRCm38 | |

LOC101055656
Genomic Map