siRNA Details
| Gene Symbol: 5530601H04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144934.1 | - | 3, 4 | 230 | ||
| AK153718.1 | - | 168 | ||||
| BC019699.1 | - | 171 | ||||
| BC055463.1 | - | 3, 4 | 203 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5530601H04Rik| Gene Symbol: 5530601H04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144934.1 | - | 3, 4 | 230 | ||
| AK153718.1 | - | 168 | ||||
| BC019699.1 | - | 171 | ||||
| BC055463.1 | - | 3, 4 | 203 | |||