siRNA Details
| Gene Symbol: Foxp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046104.1 | - | 1 | 731 | ||
| AK052184.1 | - | 1 | 1236 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxp1| Gene Symbol: Foxp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046104.1 | - | 1 | 731 | ||
| AK052184.1 | - | 1 | 1236 | |||