siRNA Details
Gene Symbol: 9330174C13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034290.1 | - | 1 | 1013 | ||
AK053329.1 | - | 1 | 1022 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9330174C13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034290.1 | - | 1 | 1013 | ||
AK053329.1 | - | 1 | 1022 |