siRNA Details
| Gene Symbol: Gm10714 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040530.1 | 2, 3 | 170 | |||
| GenBank mRNA | AK141020.1 | - | 2, 3 | 171 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB650447.1 | - | 2, 3 | 168 | Chr. 2 - 174125394 - 174183916 [-] on Build GRCm38 | |

Gm10714
Genomic Map