siRNA Details
| Gene Symbol: 1520401O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018792.1 | - | 808 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1520401O13Rik| Gene Symbol: 1520401O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018792.1 | - | 808 | |||