siRNA Details
| Gene Symbol: B230112J18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110475.1 | 3 | 477 | |||
| GenBank mRNA | AK020970.1 | - | 3 | 478 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY727026.1 | - | 3 | 478 | Chr. 5 - 116312360 - 116318285 [+] on Build GRCm38 | |

B230112J18Rik
Genomic Map