siRNA Details
| Gene Symbol: B230112J18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110475.1 | 3 | 550 | |||
| GenBank mRNA | AK020970.1 | - | 3 | 551 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B230112J18Rik| Gene Symbol: B230112J18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110475.1 | 3 | 550 | |||
| GenBank mRNA | AK020970.1 | - | 3 | 551 | ||