siRNA Details
| Gene Symbol: Gm5127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044840.1 | - | 5 | 486 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB643050.1 | - | 5 | 486 | Chr. X - 106583186 - 106708833 [+] on Build GRCm38 | |

Gm5127
Genomic Map