siRNA Details
| Gene Symbol: Zfp770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044261.1 | - | 2 | 292 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB591488.2 | - | 2 | 289 | Chr. 2 - 114200519 - 114201438 [-] on Build GRCm38 | |

Zfp770
Genomic Map