siRNA Details
| Gene Symbol: A930040O22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020947.1 | - | 1 | 550 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY742387.1 | - | 1 | 550 | Chr. 12 - 98831813 - 98832469 [+] on Build GRCm38 | |

A930040O22Rik
Genomic Map