siRNA Details
| Gene Symbol: Tnfrsf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020946.1 | - | 487 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY742383.1 | - | 1 | 487 | Chr. 4 - 150914590 - 150915129 [+] on Build GRCm38 | |

Tnfrsf9
Genomic Map