siRNA Details
| Gene Symbol: 4833419G08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014727.1 | - | 668 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4833419G08Rik| Gene Symbol: 4833419G08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014727.1 | - | 668 | |||