siRNA Details
| Gene Symbol: Impad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003208.1 | - | 1 | 491 | ||
| AK041202.1 | - | 3192 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Impad1| Gene Symbol: Impad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003208.1 | - | 1 | 491 | ||
| AK041202.1 | - | 3192 | ||||