siRNA Details
| Gene Symbol: 1700125H03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038181.1 | 2 | 173 | |||
| NR_038189.1 | 2 | 173 | ||||
| GenBank mRNA | AK007280.1 | - | 2 | 167 | ||
| AK015741.1 | - | 2 | 167 | |||
| AK131689.1 | - | 2 | 173 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700125H03Rik| Gene Symbol: 1700125H03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038181.1 | 2 | 173 | |||
| NR_038189.1 | 2 | 173 | ||||
| GenBank mRNA | AK007280.1 | - | 2 | 167 | ||
| AK015741.1 | - | 2 | 167 | |||
| AK131689.1 | - | 2 | 173 | |||