siRNA Details
| Gene Symbol: Gm9926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040528.1 | 3 | 455 | |||
| GenBank mRNA | AK033454.1 | - | 3 | 419 | ||
| AK136461.1 | - | 3 | 456 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9926| Gene Symbol: Gm9926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040528.1 | 3 | 455 | |||
| GenBank mRNA | AK033454.1 | - | 3 | 419 | ||
| AK136461.1 | - | 3 | 456 | |||