siRNA Details
| Gene Symbol: 4930405J17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045350.1 | 2 | 257 | |||
| GenBank mRNA | AK015099.1 | - | 2 | 243 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930405J17Rik| Gene Symbol: 4930405J17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045350.1 | 2 | 257 | |||
| GenBank mRNA | AK015099.1 | - | 2 | 243 | ||