siRNA Details
| Gene Symbol: 4930405J17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045350.1 | 3 | 526 | |||
| GenBank mRNA | AK015099.1 | - | 3 | 512 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU937526.1 | - | 3 | 529 | Chr. 10 - 20199219 - 20201854 [-] on Build GRCm38 | |

4930405J17Rik
Genomic Map