siRNA Details
| Gene Symbol: 4930405A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015091.1 | - | 3 | 376 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714620.1 | - | 376 | Chr. - - [-] on Build | ||

4930405A10Rik
Genomic Map