siRNA Details
| Gene Symbol: 6330565B04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018240.1 | - | 1 | 687 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY718691.1 | - | 1 | 687 | Chr. 1 - 28570488 - 28571431 [-] on Build GRCm38 | |

6330565B04Rik
Genomic Map