siRNA Details
| Gene Symbol: 4930546C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038051.1 | 1 | 607 | |||
| GenBank mRNA | AK019744.1 | - | 1 | 608 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930546C10Rik| Gene Symbol: 4930546C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038051.1 | 1 | 607 | |||
| GenBank mRNA | AK019744.1 | - | 1 | 608 | ||