siRNA Details
| Gene Symbol: 1700086P04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005980.1 | - | 2 | 350 | ||
| AK007024.1 | - | 2 | 342 | |||
| AK019743.1 | - | 2 | 350 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700086P04Rik| Gene Symbol: 1700086P04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005980.1 | - | 2 | 350 | ||
| AK007024.1 | - | 2 | 342 | |||
| AK019743.1 | - | 2 | 350 | |||