siRNA Details
| Gene Symbol: 4930524O08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040735.1 | 6 | 1422 | |||
| GenBank mRNA | AK015891.1 | - | 6 | 1424 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930524O08Rik| Gene Symbol: 4930524O08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040735.1 | 6 | 1422 | |||
| GenBank mRNA | AK015891.1 | - | 6 | 1424 | ||