siRNA Details
Gene Symbol: 4930524O05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045316.1 | 2, 3 | 340 | |||
GenBank mRNA | AK015890.1 | - | 2, 3 | 341 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715442.1 | - | 2, 3 | 341 | Chr. 19 - 10645690 - 10648053 [-] on Build GRCm38 |
