siRNA Details
| Gene Symbol: 4930524O05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045316.1 | 3 | 369 | |||
| GenBank mRNA | AK015890.1 | - | 3 | 370 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715442.1 | - | 3 | 370 | Chr. 19 - 10645690 - 10648053 [-] on Build GRCm38 | |

4930524O05Rik
Genomic Map