siRNA Details
| Gene Symbol: 4930502C17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015676.1 | - | 1 | 752 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV264684.1 | - | 1 | 62 | Chr. 3 - 33956899 - 33957149 [+] on Build GRCm38 | |

4930502C17Rik
Genomic Map