siRNA Details
| Gene Symbol: Gm30807 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132883.1 | - | 2 | 417 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615353.1 | - | 2 | 417 | Chr. 7 - 126092493 - 126093292 [-] on Build GRCm38 | |

Gm30807
Genomic Map