siRNA Details
| Gene Symbol: Coro2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045728.1 | - | 2 | 3912 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Coro2a| Gene Symbol: Coro2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045728.1 | - | 2 | 3912 | ||