siRNA Details
| Gene Symbol: Gm13003 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040443.1 | 1 | 301 | |||
| GenBank mRNA | AK132559.1 | - | 1 | 302 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13003| Gene Symbol: Gm13003 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040443.1 | 1 | 301 | |||
| GenBank mRNA | AK132559.1 | - | 1 | 302 | ||